siRNA Details
Gene Symbol: LINC00643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015358.2 | 4 | 813 | |||
GenBank mRNA | AK096461.1 | - | 1 | 335 | ||
BX248751.1 | - | 4 | 691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
