siRNA Details
Gene Symbol: DEFB128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037732.2 | NP_001032821.1 | 2 | 197 | ||
GenBank mRNA | AF525930.1 | - | 2 | 139 | ||
BC140936.1 | - | 2 | 197 | 9056748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
