siRNA Details
Gene Symbol: OR8G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291438.1 | NP_001278367.1 | 1 | 935 | ||
GenBank mRNA | BC136638.1 | - | 1 | 935 | 9020628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
