siRNA Details
Gene Symbol: LOC102724562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135824.1 | 4 | 661 | |||
GenBank mRNA | AK307907.1 | - | 2 | 844 | ||
AK311553.1 | - | 1 | 1085 | |||
CN274502.1 | - | 3 | 82 |
Gene Symbol: MST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC044862.1 | - | 6 | 3152 |
Gene Symbol: MST1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271733.1 | NP_001258662.1 | 10 | 1219 | ||
GenBank mRNA | AY192149.1 | - | 10 | 1219 |
Gene Symbol: MST1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027504.1 | 7 | 1445 | |||
GenBank mRNA | AK125231.1 | - | 2 | 870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
