siRNA Details
Gene Symbol: ERCC6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017669.2 | NP_060139.2 | 2 | 907 | ||
GenBank mRNA | AK056494.1 | - | 3 | 1044 | ||
AK074719.1 | - | 1 | 381 | |||
BC111486.1 | - | 2 | 830 | 5529990 | ||
EU069463.1 | - | 2 | 810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
