siRNA Details
Gene Symbol: NUDT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012758.2 | NP_001012776.1 | 2 | 325 | ||
XM_011509262.1 | XP_011507564.1 | 2 | 973 | |||
XM_017000560.1 | XP_016856049.1 | 2 | 973 | |||
GenBank mRNA | AK091752.1 | - | 1 | 1531 | ||
AK298083.1 | - | 2 | 326 | |||
BC046352.1 | - | 1 | 33 | |||
BG820863.1 | - | 2 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
