siRNA Details
Gene Symbol: NUDT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012758.2 | NP_001012776.1 | 8 | 911 | ||
XM_011509260.2 | XP_011507562.1 | 6 | 1349 | |||
GenBank mRNA | AA287243.1 | - | 3 | 243 | ||
AK091752.1 | - | 4 | 2170 | |||
BC046352.1 | - | 6 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
