siRNA Details
Gene Symbol: ZNF880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145434.1 | NP_001138906.1 | 4 | 1473 | ||
XM_017026812.1 | XP_016882301.1 | 4 | 1490 | |||
GenBank mRNA | AK001357.1 | - | 1505 | |||
AK297834.1 | - | 4 | 1439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
