siRNA Details
Gene Symbol: FAM201B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135203.1 | 1 | 1236 | |||
GenBank mRNA | BC045732.1 | - | 1 | 1212 |
Gene Symbol: LOC100132249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135128.1 | 1 | 1187 | |||
GenBank mRNA | AB593083.1 | - | 1 | 1188 |
Gene Symbol: LOC107987363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030106.1 | XP_016885595.1 | 1 | 1068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
