siRNA Details
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034845.2 | NP_001030017.2 | 5 | 1193 | ||
XM_011531993.2 | XP_011530295.1 | 4 | 531 | |||
XM_017008243.1 | XP_016863732.1 | 5 | 1927 | |||
XM_017008244.1 | XP_016863733.1 | 4 | 1299 | |||
GenBank mRNA | AJ626725.1 | - | 4 | 406 | ||
AK295464.1 | - | 4 | 610 | |||
BC047551.1 | - | 5 | 1114 | 5301536 | ||
GU220060.1 | - | 4 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
