siRNA Details
Gene Symbol: SERPINA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080451.1 | NP_001073920.1 | 5 | 1421 | ||
XM_006720105.3 | XP_006720168.1 | 5 | 976 | |||
GenBank mRNA | BX248259.1 | - | 5 | 1417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
