siRNA Details
Gene Symbol: PLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243079.1 | NP_001230008.1 | 12 | 1341 | ||
GenBank mRNA | AK054808.1 | - | 12 | 1341 | ||
AK122648.1 | - | 11 | 1513 | |||
BC100791.2 | - | 9 | 905 | |||
BC100792.1 | - | 9 | 905 | |||
DQ424898.1 | - | 6 | 625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
