siRNA Details
Gene Symbol: C2orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017927.3 | NP_001017927.2 | 3 | 647 | ||
NM_001322329.1 | NP_001309258.1 | 3 | 568 | |||
NM_001322330.1 | NP_001309259.1 | 2 | 444 | |||
NM_001322331.1 | NP_001309260.1 | 2 | 97 | |||
NM_001322332.1 | NP_001309261.1 | 2 | 97 | |||
XM_017003354.1 | XP_016858843.1 | 2 | 146 | |||
GenBank mRNA | BC047365.1 | - | 3 | 568 | 5189916 | |
BC062434.1 | - | 1 | 79 | |||
BC105805.1 | - | 2 | 101 | 6496685 | ||
BC126395.1 | - | 2 | 384 | 8992111 | ||
BC126397.1 | - | 3 | 587 | 8992113 | ||
BC144012.1 | - | 3 | 587 | 9052531 | ||
BC171737.1 | - | 2 | 384 | 9052532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
