siRNA Details
Gene Symbol: C2orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017927.3 | NP_001017927.2 | 7 | 1081 | ||
NM_001322329.1 | NP_001309258.1 | 7 | 1002 | |||
NM_001322330.1 | NP_001309259.1 | 6 | 878 | |||
NM_001322331.1 | NP_001309260.1 | 6 | 531 | |||
NM_001322332.1 | NP_001309261.1 | 5 | 480 | |||
GenBank mRNA | BC047365.1 | - | 7 | 1002 | 5189916 | |
BC062434.1 | - | 5 | 513 | |||
BC105805.1 | - | 6 | 535 | 6496685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
