siRNA Details
Gene Symbol: TMEM262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242631.2 | NP_001229560.1 | 1 | 187 | ||
NM_001282448.1 | NP_001269377.1 | 1 | 187 | |||
GenBank mRNA | AK124141.1 | - | 1 | 185 | ||
BX093670.1 | - | 1 | 142 | |||
HY032040.1 | - | 1 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
