siRNA Details
Gene Symbol: DEFB131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040448.2 | NP_001035538.2 | 2 | 79 | ||
GenBank mRNA | AF529414.1 | - | 2 | 79 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
