siRNA Details
Gene Symbol: MEI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282136.2 | NP_001269065.1 | 3 | 766 | ||
NM_001322247.1 | NP_001309176.1 | 3 | 371 | |||
XM_005248774.3 | XP_005248831.1 | 2 | 144 | |||
XM_017010155.1 | XP_016865644.1 | 3 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
