siRNA Details
Gene Symbol: RTEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283009.1 | NP_001269938.1 | 26 | 3195 | ||
NM_001283010.1 | NP_001269939.1 | 25 | 2916 | |||
NM_016434.3 | NP_057518.1 | 26 | 3195 | |||
NM_032957.4 | NP_116575.3 | 26 | 3267 | |||
GenBank mRNA | AF217795.1 | - | 26 | 3195 | ||
AK000485.1 | - | 2 | 398 | |||
AK299332.1 | - | 26 | 2690 | |||
AK302508.1 | - | 26 | 2786 | |||
AK304798.1 | - | 25 | 2412 | |||
AL080127.1 | - | 3 | 248 | |||
JX119186.1 | - | 25 | 2368 |
Gene Symbol: RTEL1-TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037882.1 | 26 | 3195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
