siRNA Details
Gene Symbol: MAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019090.2 | NP_061963.2 | 1 | 3052 | ||
GenBank mRNA | AB037804.1 | - | 1 | 2501 | ||
AK290216.1 | - | 1 | 2578 | |||
BC036663.2 | - | 1 | 2530 | 5266476 | ||
BC109122.2 | - | 1 | 2918 | 40036000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
