siRNA Details
Gene Symbol: MST1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271733.1 | NP_001258662.1 | 15 | 3168 | ||
GenBank mRNA | AY192149.1 | - | 15 | 3168 | ||
U28055.1 | - | 8 | 1959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
