siRNA Details
Gene Symbol: KTN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027123.1 | 3 | 607 | |||
GenBank mRNA | BX248296.1 | - | 3 | 589 | ||
JX046911.1 | - | 3 | 612 | |||
JX628584.1 | - | 3 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
