siRNA Details
Gene Symbol: LINC01551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026731.1 | 3 | 1156 | |||
GenBank mRNA | BX248251.1 | - | 3 | 1063 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
