siRNA Details
Gene Symbol: ISPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101417.3 | NP_001094887.1 | 2 | 270 | ||
NM_001101426.3 | NP_001094896.1 | 2 | 270 | |||
XM_006715770.3 | XP_006715833.1 | 2 | 440 | |||
XM_011515499.2 | XP_011513801.1 | 2 | 415 | |||
XM_011515500.2 | XP_011513802.1 | 2 | 415 | |||
XM_011515501.2 | XP_011513803.1 | 2 | 443 | |||
XM_011515508.2 | XP_011513810.1 | 2 | 772 | |||
XM_011515509.2 | XP_011513811.1 | 2 | 1621 | |||
XM_017012575.1 | XP_016868064.1 | 2 | 415 | |||
XM_017012576.1 | XP_016868065.1 | 2 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
