siRNA Details
Gene Symbol: FAM183A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101376.2 | NP_001094846.2 | 1 | 139 | ||
XM_005270875.4 | XP_005270932.1 | 1 | 968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
