siRNA Details
Gene Symbol: OR1J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014623.1 | XP_016870112.1 | 10 | 3202 | ||
GenBank mRNA | AK123016.1 | - | 4 | 2078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
