siRNA Details
Gene Symbol: TRABD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080824.2 | NP_001074293.1 | 2 | 496 | ||
NM_001277053.1 | NP_001263982.1 | 2 | 496 | |||
NM_001307978.1 | NP_001294907.1 | 2 | 496 | |||
XM_011532504.1 | XP_011530806.1 | 2 | 296 | |||
GenBank mRNA | AK296608.1 | - | 2 | 333 | ||
AK309615.1 | - | 2 | 335 | |||
BC051789.1 | - | 2 | 497 | |||
JQ653415.1 | - | 2 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
