siRNA Details
Gene Symbol: CCT6P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003110.2 | 3 | 408 | |||
GenBank mRNA | BC052238.1 | - | 3 | 408 | ||
BC073761.1 | - | 3 | 408 |
Gene Symbol: CCT6P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033416.1 | 3 | 447 | |||
GenBank mRNA | BX649060.1 | - | 3 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
