siRNA Details
Gene Symbol: VSTM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146339.1 | NP_001139811.1 | 2 | 114 | ||
XM_011526902.2 | XP_011525204.1 | 3 | 734 | |||
XM_011526903.2 | XP_011525205.1 | 3 | 734 | |||
XM_011526904.2 | XP_011525206.1 | 2 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
