siRNA Details
Gene Symbol: KIF18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001264573.1 | NP_001251503.1 | 15 | 4088 | ||
NM_001265577.1 | NP_001252506.1 | 16 | 3825 | |||
XM_011524385.2 | XP_011522687.1 | 16 | 4162 | |||
XM_011524386.2 | XP_011522688.1 | 16 | 3896 | |||
XM_011524387.2 | XP_011522689.1 | 16 | 4136 | |||
XM_011524388.2 | XP_011522690.1 | 16 | 4126 | |||
XM_011524389.2 | XP_011522691.1 | 15 | 4389 | |||
XM_011524390.2 | XP_011522692.1 | 15 | 4058 | |||
GenBank mRNA | BC044933.1 | - | 6 | 2721 | ||
BC067365.1 | - | 3 | 2250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
