siRNA Details
Gene Symbol: KIF18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001264573.1 | NP_001251503.1 | 12 | 1745 | ||
NM_001265577.1 | NP_001252506.1 | 12 | 1709 | |||
XM_011524385.2 | XP_011522687.1 | 12 | 2046 | |||
XM_011524386.2 | XP_011522688.1 | 12 | 1780 | |||
XM_011524387.2 | XP_011522689.1 | 12 | 2047 | |||
XM_011524388.2 | XP_011522690.1 | 12 | 2010 | |||
XM_011524389.2 | XP_011522691.1 | 12 | 2046 | |||
XM_011524390.2 | XP_011522692.1 | 11 | 1942 | |||
GenBank mRNA | BC044933.1 | - | 3 | 378 | ||
BC136590.1 | - | 11 | 1635 | 9020580 | ||
BC144271.1 | - | 11 | 1671 | 9052791 | ||
GU808359.1 | - | 11 | 1674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
