siRNA Details
Gene Symbol: CFAP73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144872.1 | NP_001138344.1 | 3 | 242 | ||
XM_011538327.2 | XP_011536629.1 | 3 | 938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
