siRNA Details
Gene Symbol: VSIG10L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163922.1 | NP_001157394.1 | 10 | 3108 | ||
GenBank mRNA | BC016993.1 | - | 7 | 1727 | ||
BC032383.1 | - | 7 | 1721 | |||
BC049212.1 | - | 7 | 1774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
