siRNA Details
Gene Symbol: SGSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039948.3 | NP_001035037.1 | 4, 5 | 412 | ||
NM_001098497.2 | NP_001091967.1 | 4, 5 | 412 | |||
NM_001098498.2 | NP_001091968.1 | 4, 5 | 412 | |||
NM_133454.3 | NP_597711.1 | 4, 5 | 412 | |||
GenBank mRNA | AB075821.1 | - | 1681 | |||
AB275761.1 | - | 4, 5 | 412 | |||
AB449879.1 | - | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
