siRNA Details
Gene Symbol: SLC25A30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010875.3 | NP_001010875.1 | 10 | 1396 | ||
NM_001286806.1 | NP_001273735.1 | 9 | 1277 | |||
NM_001286807.1 | NP_001273736.1 | 10 | 1454 | |||
XM_005266321.2 | XP_005266378.1 | 9 | 1544 | |||
XM_011535027.2 | XP_011533329.1 | 9 | 1173 | |||
XM_017020524.1 | XP_016876013.1 | 10 | 1446 | |||
GenBank mRNA | AK094133.1 | - | 10 | 1323 | ||
AK095465.1 | - | 9 | 1176 | |||
AK302967.1 | - | 9 | 1204 | |||
AL832206.1 | - | 4 | 3694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
