siRNA Details
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 23 | 2895 | ||
NM_173636.4 | NP_775907.4 | 23 | 2895 | |||
XM_005258809.2 | XP_005258866.1 | 23 | 2895 | |||
XM_011526837.1 | XP_011525139.1 | 23 | 2880 | |||
XM_011526838.1 | XP_011525140.1 | 23 | 2895 | |||
XM_011526839.1 | XP_011525141.1 | 21 | 2544 | |||
XM_011526840.2 | XP_011525142.1 | 16 | 1890 | |||
XM_011526841.2 | XP_011525143.1 | 14 | 1511 | |||
XM_011526842.1 | XP_011525144.1 | 13 | 1343 | |||
XM_011526843.1 | XP_011525145.1 | 6 | 670 | |||
XM_011526844.2 | XP_011525146.1 | 6 | 667 | |||
XM_017026665.1 | XP_016882154.1 | 23 | 2895 | |||
GenBank mRNA | AL133651.1 | - | 2 | 212 | ||
BC017261.2 | - | 5 | 604 | |||
BC058939.1 | - | 23 | 2839 | |||
BX647726.1 | - | 24 | 2956 | |||
KU178897.1 | - | 2 | 140 | |||
KU178898.1 | - | 2 | 140 | |||
KU178899.1 | - | 2 | 140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
