siRNA Details
Gene Symbol: CEP170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544338.2 | XP_011542640.1 | 14, 15 | 5133 | ||
XM_011544342.2 | XP_011542644.1 | 14, 15 | 4975 | |||
XM_017002935.1 | XP_016858424.1 | 15, 16 | 4897 | |||
XM_017002939.1 | XP_016858428.1 | 15, 16 | 4717 | |||
XM_017002940.1 | XP_016858429.1 | 15, 16 | 4360 | |||
XM_017002942.1 | XP_016858431.1 | 15, 16 | 4789 | |||
XM_017002943.1 | XP_016858432.1 | 15, 16 | 4609 | |||
XM_017002945.1 | XP_016858434.1 | 14, 15 | 4653 | |||
XM_017002949.1 | XP_016858438.1 | 14, 15 | 4545 | |||
XM_017002950.1 | XP_016858439.1 | 14, 15 | 4420 |
Gene Symbol: CEP170P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003135.2 | 3, 4 | 271 | |||
GenBank mRNA | BC014590.2 | - | 3, 4 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
