siRNA Details
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009909.3 | NP_001009909.2 | 12 | 1473 | ||
NM_001252008.1 | NP_001238937.1 | 12 | 1469 | |||
NM_001252010.1 | NP_001238939.1 | 10 | 1347 | |||
GenBank mRNA | AK126974.1 | - | 4 | 516 | ||
AK309077.1 | - | 10 | 1122 | |||
AL832641.3 | - | 12 | 1385 | |||
BC132829.1 | - | 12 | 1265 | 40146851 | ||
BC144177.1 | - | 12 | 1261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
