siRNA Details
Gene Symbol: CDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029954.2 | NP_001025125.2 | 2 | 185 | ||
XM_011519488.2 | XP_011517790.1 | 2 | 274 | |||
GenBank mRNA | AV657190.1 | - | 2 | 281 | ||
BC037872.1 | - | 4 | 350 | |||
BC133042.1 | - | 2 | 172 | 40147064 | ||
BC133044.1 | - | 2 | 172 | 40147066 | ||
BC144619.1 | - | 2 | 172 | |||
CN311321.1 | - | 2 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
