siRNA Details
Gene Symbol: VSIG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013661.1 | NP_001013683.1 | 3 | 392 | ||
GenBank mRNA | BC132893.1 | - | 3 | 284 | 40146915 | |
BC132895.1 | - | 3 | 284 | 40146917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
