siRNA Details
Gene Symbol: NDOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144026.2 | NP_001137498.1 | 14 | 4405 | ||
NM_001144027.2 | NP_001137499.1 | 13 | 4254 | |||
NM_001144028.2 | NP_001137500.1 | 14 | 4357 | |||
NM_014434.3 | NP_055249.1 | 14 | 4378 | |||
XM_011518545.1 | XP_011516847.1 | 13 | 4293 | |||
XM_011518547.1 | XP_011516849.1 | 14 | 4195 | |||
XM_011518548.2 | XP_011516850.1 | 13 | 5595 | |||
XM_017014627.1 | XP_016870116.1 | 14 | 4356 | |||
GenBank mRNA | AL133632.1 | - | 1 | 830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
