siRNA Details
Gene Symbol: CCDC157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017437.3 | NP_001017437.2 | 5, 6 | 1718 | ||
NM_001318334.1 | NP_001305263.1 | 5, 6 | 1764 | |||
XM_006724278.3 | XP_006724341.1 | 5, 6 | 1325 | |||
XM_006724279.3 | XP_006724342.1 | 5, 6 | 1325 | |||
XM_006724280.3 | XP_006724343.1 | 5, 6 | 1325 | |||
XM_011530263.2 | XP_011528565.1 | 5, 6 | 1378 | |||
XM_011530266.2 | XP_011528568.1 | 5, 6 | 1325 | |||
XM_011530267.2 | XP_011528569.1 | 5, 6 | 1325 | |||
XM_011530268.2 | XP_011528570.1 | 5, 6 | 1325 | |||
XM_011530269.2 | XP_011528571.1 | 1, 2 | 95 | |||
XM_011530270.2 | XP_011528572.1 | 5, 6 | 1325 | |||
GenBank mRNA | BC092419.1 | - | 5, 6 | 1352 | 30383661 | |
CR936833.1 | - | 4 | 1679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
