siRNA Details
Gene Symbol: CCDC157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017437.3 | NP_001017437.2 | 10 | 2484 | ||
NM_001318334.1 | NP_001305263.1 | 10 | 2530 | |||
XM_006724278.3 | XP_006724341.1 | 10 | 2112 | |||
XM_006724279.3 | XP_006724342.1 | 10 | 2046 | |||
XM_011530263.2 | XP_011528565.1 | 10 | 2165 | |||
XM_011530269.2 | XP_011528571.1 | 6 | 882 | |||
GenBank mRNA | BC092419.1 | - | 10 | 2118 | 30383661 | |
BC119799.1 | - | 2 | 228 | |||
BC127255.1 | - | 2 | 228 | |||
CR936833.1 | - | 8 | 2438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
