siRNA Details
Gene Symbol: EFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024768.2 | NP_079044.2 | 1 | 628 | ||
XM_005247781.3 | XP_005247838.1 | 1 | 653 | |||
XM_011513163.2 | XP_011511465.1 | 1 | 653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
