siRNA Details
Gene Symbol: MFSD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080473.1 | NP_001073942.1 | 11 | 1154 | ||
XM_005264301.4 | XP_005264358.1 | 11 | 2215 | |||
XM_017004063.1 | XP_016859552.1 | 10 | 2308 | |||
XM_017004064.1 | XP_016859553.1 | 10 | 2293 | |||
XM_017004065.1 | XP_016859554.1 | 9 | 2224 | |||
GenBank mRNA | BX647146.1 | - | 6 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
