siRNA Details
Gene Symbol: MFSD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080473.1 | NP_001073942.1 | 12 | 1257 | ||
XM_005264301.4 | XP_005264358.1 | 12 | 2318 | |||
XM_017004063.1 | XP_016859552.1 | 11 | 2411 | |||
XM_017004064.1 | XP_016859553.1 | 11 | 2396 | |||
XM_017004065.1 | XP_016859554.1 | 10 | 2327 | |||
XM_017004066.1 | XP_016859555.1 | 11 | 2379 | |||
XM_017004067.1 | XP_016859556.1 | 10 | 2301 | |||
XM_017004068.1 | XP_016859557.1 | 8 | 2136 | |||
GenBank mRNA | BX647146.1 | - | 6 | 1022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
