siRNA Details
Gene Symbol: ZNF847P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036753.1 | 1 | 69 | |||
GenBank mRNA | AF387613.1 | - | 1 | 69 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
