siRNA Details
Gene Symbol: FAM227A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013647.1 | NP_001013669.1 | 7, 8 | 1224 | ||
NM_001291030.1 | NP_001277959.1 | 7, 8 | 1264 | |||
XM_006724291.2 | XP_006724354.2 | 7, 8 | 1224 | |||
XM_006724292.2 | XP_006724355.2 | 7, 8 | 1224 | |||
XM_006724293.2 | XP_006724356.1 | 7, 8 | 1224 | |||
XM_006724294.2 | XP_006724357.2 | 7, 8 | 1264 | |||
XM_006724295.2 | XP_006724358.2 | 7, 8 | 1264 | |||
XM_011530308.2 | XP_011528610.1 | 7, 8 | 929 | |||
XM_011530309.2 | XP_011528611.1 | 7, 8 | 1224 | |||
XM_011530311.1 | XP_011528613.1 | 6, 7 | 1101 | |||
XM_011530312.1 | XP_011528614.1 | 5, 6 | 501 | |||
XM_011530313.2 | XP_011528615.1 | 5, 6 | 453 | |||
XM_011530314.1 | XP_011528616.1 | 6, 7 | 1154 | |||
GenBank mRNA | AK301819.1 | - | 7, 8 | 938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
