siRNA Details
Gene Symbol: FAM227A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013647.1 | NP_001013669.1 | 2 | 703 | ||
NM_001291030.1 | NP_001277959.1 | 2 | 703 | |||
XM_006724291.2 | XP_006724354.2 | 2 | 703 | |||
XM_006724292.2 | XP_006724355.2 | 2 | 703 | |||
XM_006724293.2 | XP_006724356.1 | 2 | 703 | |||
XM_006724294.2 | XP_006724357.2 | 2 | 703 | |||
XM_006724295.2 | XP_006724358.2 | 2 | 703 | |||
XM_011530308.2 | XP_011528610.1 | 2 | 408 | |||
XM_011530309.2 | XP_011528611.1 | 2 | 703 | |||
XM_011530310.1 | XP_011528612.1 | 2 | 703 | |||
XM_011530314.1 | XP_011528616.1 | 2 | 703 | |||
GenBank mRNA | AK301819.1 | - | 2 | 377 | ||
DB053353.1 | - | 2 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
