siRNA Details
Gene Symbol: UBE2QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145161.2 | NP_001138633.1 | 2 | 1361 | ||
GenBank mRNA | AB188494.1 | - | 2 | 1734 | ||
AK057805.1 | - | 2 | 832 | |||
AK096612.1 | - | 1 | 396 | |||
AK308577.1 | - | 2 | 1125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
