siRNA Details
Gene Symbol: SPRED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042522.2 | NP_001035987.1 | 4 | 611 | ||
NR_073032.1 | 5 | 542 | ||||
XM_006723219.3 | XP_006723282.1 | 5 | 602 | |||
XM_011526978.2 | XP_011525280.1 | 4 | 657 | |||
GenBank mRNA | BC151136.1 | - | 4 | 611 | 9057036 | |
BF983829.1 | - | 5 | 543 | |||
DQ323928.1 | - | 4 | 512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
